Learn More
Enzo Life Sciences Arg8-Vasopressin ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900017A
Encompass_Preferred
Highly sensitive ELISA kit for the detection of Arg8-Vasopressin in serum, plasma and tissue culture media. The Arg8-Vasopressin EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour. Absorbance is read at 450 nm. Alternative Name: AVP. Sensitivity: 2.84 pg/ml (range 4.10 - 1,000 pg/ml). Assay Time: Overnight + 1 hour. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, SA-HRP, TMB Substrate, Stop solution
Catalog No. NC1073905