Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 383MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: ARG1/1126. Clonality: Monoclonal. Host: Mouse. Isotype: IgG3 kappa. Positive Control: 293T cells. Hepatocellular Carcinoma (HCC). Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant fragment (87 Amino acid residues around aa 1-150) of human ARG1 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 35-38kDa.Chromosome: 6q23. Gene Symbol: ARG1. Gene ID: 383. Ensemble: ENSG00000118520. Uniprot: XP_011534103.
Catalog No. 50-188-4852