Learn More
Aviva Systems Biology Corp ARG1 Antibody - N-terminal region (ARP45671_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45671_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARG1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ARG1. Alternate names: -ApplicationWB, IHC. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-59095