Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Arg1-in-1 (hydrochloride) | 00-00-0 | 90.0% | 292.57 g·mol⁻1 | C11H22BClN2O4 | 5 MG
SDP

Catalog No. 5000221232
Encompass_Preferred

ARG1-IN-1 hydrochloride is a small-molecule inhibitor of human arginase 1 with a reported IC50 of 29 nM. It is intended for research use in biochemical and cellular studies to probe arginase-related pathways. The hydrochloride form improves handling and aqueous compatibility for assay preparation.

  • Potent arginase 1 inhibition (IC50 29 nM).
  • Suitable for biochemical and cellular assays.
  • Provided as a 5 mg quantity for small-scale research use.
  • Molecular weight 292.57 g·mol⁻1; formula C11H22BClN2O4.
  • Purity typically 90.0% as reported on the certificate of analysis.
  • Store at -20°C under nitrogen and protect from moisture; in solvent store at -80°C for long-term.

Catalog No. 50-002-21232 Supplier Medchemexpress LLC Supplier No. HY145331A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ARG1-IN-1 hydrochloride is a small-molecule inhibitor of human arginase 1 with a reported IC50 of 29 nM. It is intended for research use in biochemical and cellular studies to probe arginase-related pathways. The hydrochloride form improves handling and aqueous compatibility for assay preparation.

  • Potent arginase 1 inhibition (IC50 29 nM).
  • Suitable for biochemical and cellular assays.
  • Provided as a 5 mg quantity for small-scale research use.
  • Molecular weight 292.57 g·mol⁻1; formula C11H22BClN2O4.
  • Purity typically 90.0% as reported on the certificate of analysis.
  • Store at -20°C under nitrogen and protect from moisture; in solvent store at -80°C for long-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.