Learn More
Medchemexpress LLC ARG1-IN-1 hydrochloride | 00-00-0 | 90.0% | C11H22BClN2O4 | 10MG
SDP
Catalog No. 5000259123
Encompass_Preferred
ARG1-IN-1 hydrochloride is a research compound that selectively inhibits human arginase 1, with a reported IC50 of 29 nM. Supplied as the hydrochloride salt, the material is typically offered at 90.0% purity and is intended for biochemical and pharmacological research applications only.
  • Potent arginase 1 inhibition (IC50 = 29 nM).
  • Supplied as a hydrochloride salt for chemical stability.
  • Typical purity 90.0% suitable for research use.
  • Appropriate for in vitro biochemical and cell-based assays.
  • Provided in milligram-scale quantities for laboratory studies.
Catalog No. 50-002-59123 Supplier Medchemexpress LLC Supplier No. HY145331A10MG
ARG1-IN-1 hydrochloride is a research compound that selectively inhibits human arginase 1, with a reported IC50 of 29 nM. Supplied as the hydrochloride salt, the material is typically offered at 90.0% purity and is intended for biochemical and pharmacological research applications only.
  • Potent arginase 1 inhibition (IC50 = 29 nM).
  • Supplied as a hydrochloride salt for chemical stability.
  • Typical purity 90.0% suitable for research use.
  • Appropriate for in vitro biochemical and cell-based assays.
  • Provided in milligram-scale quantities for laboratory studies.