Learn More
Medchemexpress LLC Arg1-in-1 hydrochloride | 90.0% | C11H22BClN2O4 | 1 MG
SDP
Catalog No. 5000354907
Encompass_Preferred
ARG1-IN-1 hydrochloride is an orally active arginase inhibitor with an IC50 value of 29 nM. It can inhibit serum arginase activity and increase arginine levels in mouse tumor models, making it applicable for tumor research.
  • Orally active arginase inhibitor.
  • Inhibits serum arginase activity.
  • Increases arginine levels in mouse tumor models.
  • Applicable for tumor research.
Catalog No. 50-003-54907 Supplier Medchemexpress LLC Supplier No. HY145331A1MG
ARG1-IN-1 hydrochloride is an orally active arginase inhibitor with an IC50 value of 29 nM. It can inhibit serum arginase activity and increase arginine levels in mouse tumor models, making it applicable for tumor research.
  • Orally active arginase inhibitor.
  • Inhibits serum arginase activity.
  • Increases arginine levels in mouse tumor models.
  • Applicable for tumor research.