Learn More
Felix Storch Inc ACCUCOLD 18 Cu.Ft. Upright Vaccine Refrigerator, Certified to NSF/ANSI 456 Vaccine Storage Standard
SDP
Supplier:  Felix Storch Inc ARG18PV456
Encompass_Preferred
ARG18PV456 18CF GLASS DOOR IS CERTIFIED PURPSOE BUILT for SAFE VACCINE STORAGE. Accucold is proud to lead the industry with the largest line of cold storage product sizes certified to NSF/ANSI 456 Vaccine Storage Standard. The Pharma-Vac Performance Series supports meeting regulations and guidelines from the CDC, State Agencies, World Health Organization, and other Health Ministries on vaccine storage and handling. Temp. Security and Saftey Features: https://www.accucold.com/product/ARG18PV456
Catalog No. NC2260534
Add to Cart
Request a Quote