Learn More
Aviva Systems Biology Corp ARG2 Antibody - N-terminal region (ARP54566_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54566_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARG2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ARG2. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-940