Learn More
CPC Scientific Ac-Ser-Gly-Arg(Me2)-Gly-Lys-Gly-Gly-Lys-Gly-Leu-Gly-Lys-Gly-Gly-Ala-Lys-Arg-His-Arg-Lys-Val-Gly-Gly-Lys(Biotin)-OH (trifluoroacetate salt) 0.25MG
SDP
Supplier:  CPC Scientific HIST077A
Encompass
This 21-mer peptide is a member of the Histone H4 category and contains a dimethylated (symmetrically) arginine (Arg3) and a C-terminal Gly-Gly linker followed by a biotinylated lysine.ONE-LETTER SEQUENCE: Ac-SG-R(me2s)-GKGGKGLGKGGAKRHRKV-GGK(Biotin)MOLECULAR FORMULA: MOLECULAR WEIGHT:2630.1STORAGE CONDITIONS: -20 5CSYNONYMS: H4R3(Me2s) (1-21), biotin - labeledRESEARCH AREA: Miscellaneous
Catalog No. 50-210-9518