Learn More
Felix Storch Inc ACCUCOLD 2.83 Cu.Ft. ADA Height Vaccine Refrigerator, Certified to NSF/ANSI 456 Vaccine Storage Standard
SDP
Catalog No. NC2260515
Encompass_Preferred
ARG31PVBIADA456 2.83CF GLASS DOOR, BUILT-IN, ADA HEIGHT IS CERTIFIED PURPSOE BUILT for SAFE VACCINE STORAGE. Accucold is proud to lead the industry with the largest line of cold storage product sizes certified to NSF/ANSI 456 Vaccine Storage Standard. The Pharma-Vac Performance Series supports meeting regulations and guidelines from the CDC, State Agencies, World Health Organization, and other Health Ministries on vaccine storage and handling. Temp. Security and Saftey Features: https://www.accucold.com/product/ARG31PVBIADA456
Catalog No. NC2260515 Supplier Felix Storch Inc Supplier No. ARG31PVBIADA456
ARG31PVBIADA456 2.83CF GLASS DOOR, BUILT-IN, ADA HEIGHT IS CERTIFIED PURPSOE BUILT for SAFE VACCINE STORAGE. Accucold is proud to lead the industry with the largest line of cold storage product sizes certified to NSF/ANSI 456 Vaccine Storage Standard. The Pharma-Vac Performance Series supports meeting regulations and guidelines from the CDC, State Agencies, World Health Organization, and other Health Ministries on vaccine storage and handling. Temp. Security and Saftey Features: https://www.accucold.com/product/ARG31PVBIADA456