Learn More
Bachem (Arg8)-Vasopressin Trifluoroacetate
SDP
Supplier:  Bachem 4012215.0001
Encompass_Preferred
(Arg8)-Vasopressin Trifluoroacetate, H-Cys-Tyr-Phe-Gln-Asn-Cys-Pro-Arg-Gly-NH2, 113-79-1, C46H65N15O12S2, Mr 1084.25, 1mg. Argipressin, AVP, (Phe3,Arg8)-Oxytocin, Leiormone, Arginine Antidiuretic Hormone. The major physiological roles of Arg-Vasopressin are regulation of water balance in animals (antidiuretic action) and contraction of arterioles (vasopressor action). CAS number (argipressin acetate): 129979-57-3.
Catalog No. 50-259-538