Learn More
Arbor Assays Arg8-Vasopressin (AVP) ELISA Kit (1 Plate)
SDP
Supplier:  Arbor Assays K049H1
Encompass_Preferred
The DetectX Arg8-Vasopressin (AVP) Immunoassay Kit (ELISA) is designed to quantitatively measure AVP present in serum, plasma and tissue culture media samples.
Catalog No. 50-227-8465