Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Arg8-Vasopressin Competitive ELISA Kit

Catalog No. EIAAVP
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIAAVP 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EIAAVP Supplier Invitrogen™ Supplier No. EIAAVP
Add to Cart
Add to Cart

ELISA

The multispecies Arg8-Vasopressin (AVP) Competivite ELISA with quantitates Arg8-Vasopressin in serum, plasma, and tissue culture media. Principle of the method: The Arg8-Vasopressin (AVP) Competitive ELISA research-use-only kit is designed to quantitatively measure AVP independent of species. An AVP standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with goat anti-rabbit IgG antibodies. Rabbit polyclonal antibodies and AVP-peroxidase conjugate is added to the wells. The AVP conjugate and any AVP in the sample will compete to bind to the rabbit polyclonal AVP antibodies. After 30 min incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm. The intensity of the generated color corresponds inversely to the amount of AVP present in the sample. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

This gene encodes a precursor protein consisting of arginine vasopressin and two associated proteins, neurophysin 2 and a glycopeptide, copeptin. Arginine vasopressin is a posterior pituitary hormone which is synthesized in the supraoptic nucleus and paraventricular nucleus of the hypothalamus. Along with its carrier protein, neurophysin 2, it is packaged into neurosecretory vesicles and transported axonally to the nerve endings in the neurohypophysis where it is either stored or secreted into the bloodstream. The precursor is thought to be activated while it is being transported along the axon to the posterior pituitary. Arginine vasopressin acts as a growth factor by enhancing pH regulation through acid-base transport systems. It has a direct antidiuretic action on the kidney, and also causes vasoconstriction of the peripheral vessels. This hormone can contract smooth muscle during parturition and lactation. It is also involved in cognition, tolerance, adaptation and complex sexual and maternal behaviour, as well as in the regulation of water excretion and cardiovascular functions.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 4.1-1000 pg/mL
Assay Sensitivity 3.67 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Chromone
Sample Type Plasma, Serum, Tissue Homogenates
Target Species All Species
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias ADH; antidiuretic hormone; arginine vasopressin; arginine vasopressin (Diabetes insipidus), same as Di (conflicting physical mapping); arginine vasopressin (neurophysin II, antidiuretic hormone, diabetes insipidus, neurohypophyseal); arginine vasopressin-neurophysin II precursor; Arginine-vasopressin; Arg-vasopressin; ARVP; Avp; AVP-neurophysin II precursor; AVP-NPII; AVRP; Copeptin; DI; neurohypophyseal; Neurophysin 2; Neurophysin-I; Neurophysin-II; prepro-arginine-vasopressin-neurophysin II; prepro-AVP-NP II; prepropressophysin; preprovasopressin-neurophysin; Vas; Vasopressin; vasopressin-neurophysin 2-copeptin; vasopressin-neurophysin II-copeptin; vasopressin-neurophysin prepropeptide; VP; VspADH; antidiuretic hormone; arginine vasopressin; arginine vasopressin (Diabetes insipidus), same as Di (conflicting physical mapping); arginine vasopressin (neurophysin II, antidiuretic hormone, diabetes insipidus, neurohypophyseal); arginine vasopressin-neurophysin II precursor; Arginine-vasopressin; Arg-vasopressin; ARVP; Avp; AVP-neurophysin II precursor; AVP-NPII; AVRP; Copeptin; DI; neurohypophyseal; Neurophysin 2; Neurophysin-I; Neurophysin-II; prepro-arginine-vasopressin-neurophysin II; prepro-AVP-NP II; prepropressophysin; preprovasopressin-neurophysin; Vas; Vasopressin; vasopressin-neurophysin 2-copeptin; vasopressin-neurophysin II-copeptin; vasopressin-neurophysin prepropeptide; VP; Vsp;
Interassay CV <9%
Intraassay CV <9%
Kit Contents Pre-coated 96 well plate, Standard, Antibody, HRP-Conjugated Detection Antibody, Assay Buffer Concentrate, Extraction Solution, Wash Buffer, TMB Substrate, Stop Solution, Adhesive Plate Covers, Standard, Antibody, HRP-Conjugated Detection Antibody, Assay Buffer Concentrate, Wash Buffer, TMB Substrate, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 100 μL, Serum 100 μL, Tissue Homogenate 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements 2-8°C
Target AVP
Test Time 1 hr 20 min
Total Assay Time 16 hr
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.