Learn More
Enzo Life Sciences Arg8-Vasopressin ELISA kit (5x96wells)
SDP
Supplier:  Enzo Life Sciences ADI901017A
Encompass_Preferred
Highly sensitive ELISA kit for the detection of Arg8-Vasopressin in serum, plasma and tissue culture media. The Arg8-Vasopressin EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour. Absorbance is read at 450 nm. Alternative name: AVP. Sensitivity: 2.84 pg/ml (range 4.10 - 1,000 pg/ml). Use/Stability: Store all components at +4°C, except standard at -20°C. Assay Time: Overnight + 1 hour. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent .
Catalog No. 50-200-6885