Learn More
Bachem (Arg8)-Vasotocin Trifluoroacetate
SDP
Supplier:  Bachem 4031338.0005
Encompass_Preferred
(Arg8)-Vasotocin Trifluoroacetate, H-Cys-Tyr-Ile-Gln-Asn-Cys-Pro-Arg-Gly-NH2, 113-80-4, C43H67N15O12S2, Mr 1050.23, 5mg. Argiprestocin. Vasotocin (AVT, argiprestocin) is the vasopressin/oxytocin analog produced in birds, amphibians and fish. AVT shows antidiuretic as well as reproductive activities.
Catalog No. 50-259-541