Learn More
Medchemexpress LLC Arg9 TFA (Nona-L-arginine TFA) | 2283335-13-5 | 99.6% | 1423.69 (free acid) | 5 MG
SDP
Catalog No. 5000425951
Encompass_Preferred
Arg9 TFA (Nona-L-arginine) is a cell-penetrating peptide (CPP) composed of nine arginine residues. This peptide has demonstrated neuroprotective properties, showing significant activity in various models of neuronal damage. It effectively reduces cell death following exposure to excitotoxins and ischemia, making it a potential candidate for research in neurological conditions.
  • Functions as a cell-penetrating peptide (CPP).
  • Composed of nine arginine residues.
  • Exhibits neuroprotective activity with an IC50 of 0.78 μM in the glutamic acid model.
  • Provides significant neuroprotection in a dose-response manner.
  • Reduces infarct volume in a permanent middle cerebral artery stroke model in rats.
Catalog No. 50-004-25951 Supplier Medchemexpress LLC Supplier No. HYP0133A5MG
Arg9 TFA (Nona-L-arginine) is a cell-penetrating peptide (CPP) composed of nine arginine residues. This peptide has demonstrated neuroprotective properties, showing significant activity in various models of neuronal damage. It effectively reduces cell death following exposure to excitotoxins and ischemia, making it a potential candidate for research in neurological conditions.
  • Functions as a cell-penetrating peptide (CPP).
  • Composed of nine arginine residues.
  • Exhibits neuroprotective activity with an IC50 of 0.78 μM in the glutamic acid model.
  • Provides significant neuroprotection in a dose-response manner.
  • Reduces infarct volume in a permanent middle cerebral artery stroke model in rats.