Learn More
Cayman Chemical Arg9trIfluoroacetAT salt 10mg
SDP
Catalog No. 502910057
Encompass_Preferred
A cationic cell-penetrating peptide; has been used to deliver various cargo to cells, including fluorescent proteins and plasmids; protects primary rat cortical neurons from glutamate-induced excitotoxicity at 5 and 10 µM
Catalog No. 50-291-0057 Supplier Cayman Chemical Supplier No. 3671410 MG
A cationic cell-penetrating peptide; has been used to deliver various cargo to cells, including fluorescent proteins and plasmids; protects primary rat cortical neurons from glutamate-induced excitotoxicity at 5 and 10 µM