Learn More
Medchemexpress LLC Argadin | 289665-92-5 | 98.7% | 674.71 | 1 MG
SDP
Catalog No. 5000398916
Encompass_Preferred
Argadin is a cyclopentapeptide chitinase inhibitor that inhibits chitinase B (ChiB).
  • Inhibits chitinase B (ChiB) with a Ki value of 20 nM.
  • Can be used for research related to fungicides.
  • Can be used for research related to insecticides.
Catalog No. 50-003-98916 Supplier Medchemexpress LLC Supplier No. HYP34611MG
Argadin is a cyclopentapeptide chitinase inhibitor that inhibits chitinase B (ChiB).
  • Inhibits chitinase B (ChiB) with a Ki value of 20 nM.
  • Can be used for research related to fungicides.
  • Can be used for research related to insecticides.