Learn More
Medchemexpress LLC HY-B0375A 10mg Medchemexpress, Argatroban (monohydrate) CAS:141396-28-3 Purity:>98%
SDP
Catalog No. 502082499
Encompass_Preferred
Medchemexpress, HY-B0375A 10mg Argatroban (monohydrate) CAS:141396-28-3 Argatroban (monohydrate) (MD-805 (monohydrate)) is a direct, selective thrombin inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2499 Supplier Medchemexpress LLC Supplier No. HYB0375A10MG
Medchemexpress, HY-B0375A 10mg Argatroban (monohydrate) CAS:141396-28-3 Argatroban (monohydrate) (MD-805 (monohydrate)) is a direct, selective thrombin inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.