Learn More
Medchemexpress LLC Argatroban (monohydrate) | 141396-28-3 | 99.6% | 5 MG
SDP
Catalog No. 5000407125
Encompass_Preferred
Argatroban (monohydrate) is a direct, selective thrombin inhibitor used for research purposes. It is effective in preventing thrombus formation and has been studied for its use in creating animal models for thrombolysis.
  • Suitable for high-speed applications
  • Available in solid form
  • Used to inhibit thrombin activity in animals
  • Has potential for preventing thrombus formation without aggravating bleeding
  • Can enhance reperfusion with TPA in patients with AMI
Catalog No. 50-004-07125 Supplier Medchemexpress LLC Supplier No. HYB0375A5MG
Argatroban (monohydrate) is a direct, selective thrombin inhibitor used for research purposes. It is effective in preventing thrombus formation and has been studied for its use in creating animal models for thrombolysis.
  • Suitable for high-speed applications
  • Available in solid form
  • Used to inhibit thrombin activity in animals
  • Has potential for preventing thrombus formation without aggravating bleeding
  • Can enhance reperfusion with TPA in patients with AMI