Learn More
Aviva Systems Biology Corp ARGFX Antibody - N-terminal region (ARP47512_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47512_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ARGFX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ARGFX. Alternate names: -ApplicationWB. Species reactivity: Human, Yeast
Catalog No. 50-014-59574