Learn More
Cayman Chemical Arg-Gly-Asp (trifluoroacetate salt), Purity: ≥95%, CAS: 120103-89-1, 50 mg
SDP
Catalog No. 502991879
Encompass_Preferred
A synthetic peptide corresponding to an integrin cell adhesion sequence; a component of radioligands and peptidomimetics that have been used in the study of integrin expression and activity in vivo
Catalog No. 50-299-1879 Supplier Cayman Chemical Supplier No. 3136950 MG
A synthetic peptide corresponding to an integrin cell adhesion sequence; a component of radioligands and peptidomimetics that have been used in the study of integrin expression and activity in vivo