Learn More
Genetex Inc Anti-Arginase 1 antibody
SDP
Supplier:  Genetex Inc GTX109242
Encompass
The Anti-Arginase 1 antibody from GeneTex is a Rabbit Polyclonal antibody to Arginase 1. The Anti-Arginase 1 antibody Recognizes Human, Mouse, Rat. The Anti-Arginase 1 antibody has been validated for the following Applications: FACS, ICC/IF, IHC-Fr, IHC-P, IP, WB.
Catalog No. NC0934104