Learn More
Cell Signaling Technology Arginase-2 (D9J1N) Rabbit Monoclonal Antibody, Unconjugated, 100 μl
SDP
Catalog No. 502733066
Encompass
Arginase-2 (D9J1N) Rabbit Monoclonal Antibody is a recombinant antibody that recognizes endogenous levels of total arginase-2 protein. It is produced by immunizing animals with a synthetic peptide corresponding to residues near the carboxy terminus of the human arginase-2 protein. This antibody is suitable for applications including Western blotting, immunoprecipitation, immunohistochemistry (paraffin), and flow cytometry.
  • Recognizes endogenous levels of total arginase-2 protein
  • Does not cross-react with arginase-1 protein
  • Validated for use in human, mouse, and rat species
  • Recombinant monoclonal rabbit IgG
Catalog No. 50-273-3066 Supplier Cell Signaling Technology Supplier No. 55003S
Arginase-2 (D9J1N) Rabbit Monoclonal Antibody is a recombinant antibody that recognizes endogenous levels of total arginase-2 protein. It is produced by immunizing animals with a synthetic peptide corresponding to residues near the carboxy terminus of the human arginase-2 protein. This antibody is suitable for applications including Western blotting, immunoprecipitation, immunohistochemistry (paraffin), and flow cytometry.
  • Recognizes endogenous levels of total arginase-2 protein
  • Does not cross-react with arginase-1 protein
  • Validated for use in human, mouse, and rat species
  • Recombinant monoclonal rabbit IgG