Learn More
Medchemexpress LLC HY-15775 5mg Medchemexpress, Arginase inhibitor 1 CAS:1345808-25-4 Purity:>98%
SDP
Catalog No. 502081796
Encompass_Preferred
Medchemexpress, HY-15775 5mg Arginase inhibitor 1 CAS:1345808-25-4 Arginase inhibitor 1 is a potent inhibitor of human arginases I and II with IC50s of 223 and 509 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1796 Supplier Medchemexpress LLC Supplier No. HY157755MG
Medchemexpress, HY-15775 5mg Arginase inhibitor 1 CAS:1345808-25-4 Arginase inhibitor 1 is a potent inhibitor of human arginases I and II with IC50s of 223 and 509 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.