Learn More
Cayman Chemical Arginase Inhibitor 1, Purity: ≥90%, CAS: 1345808-25-4, 5 mg
SDP
Catalog No. 502939312
Encompass_Preferred
An arginase inhibitor (IC50s = 223 and 509 nM for recombinant human arginase I and II, respectively); decreases arginase I activity in CHOK cells expressing human arginase I (IC50 = 8 µM); reduces infarct size in a rat model of myocardial ischemia and reperfusion injury induced by left-main coronary artery occlusion at 100 mg/kg, i.v.
Catalog No. 50-293-9312 Supplier Cayman Chemical Supplier No. 284185 MG
An arginase inhibitor (IC50s = 223 and 509 nM for recombinant human arginase I and II, respectively); decreases arginase I activity in CHOK cells expressing human arginase I (IC50 = 8 µM); reduces infarct size in a rat model of myocardial ischemia and reperfusion injury induced by left-main coronary artery occlusion at 100 mg/kg, i.v.