Learn More
ACROBiosystems Recombinant Protein;Human Arginase 1 / ARG1 Protein;HEK293;1MG;AR1,ARG1,Arginase-1,ARG-1,Arginase 1
Supplier:  ACROBiosystems AR1H52281MG
Encompass_Preferred
Recombinant Protein;Human Arginase 1 / ARG1 Protein;1MG;Human ARG1, His Tag (AR1-H5228) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Lys 322 (Accession # NP_000036).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.6 kDa. The protein migrates as 36 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-9048