Learn More
Biomatik Corporation Arginine (Arg) CLIA Kit
Supplier:  Biomatik Corporation EKU08662
Encompass_Preferred
Arginine (Arg) CLIA Kit for detecting Arg in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Universal. Detection Range: 390.6-100,000ng/ml. Sensitivity: 146.9ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Target Synonyms: (S)-2-Amino-5-Guanidinopentanoic Acid. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8642