Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Argininosuccinic acid disodium salt hydrate

Catalog No. A570750MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A570750MG 50 mg
A570710MG 10 mg
A5707250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A570750MG Supplier Sigma Aldrich Supplier No. A570750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Argininosuccinic acid disodium salt hydrate has been used in the spectrophotometric measurement of argininosuccinate lyase (asl) enzymatic activity in cell extracts.tandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf
  • A5707.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database
  • To learn more, visit sigma.com/metlin

Specifications

CAS 918149-29-8
Molecular Formula C10H16N4Na2O6 -+ xH2O
Linear Formula C10H16N4Na2O6 -+ xH2O
Quantity 50 mg
MDL Number MFCD11656132
Molecular Weight (g/mol) 334.24 (anhydrous basis)
Percent Purity ≥80%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.