Learn More
Medchemexpress LLC Arginyl-glycyl-aspartyl-serine | 91037-65-9 | 99.1 % | 100 MG
SDP
Catalog No. 5000230960
Encompass_Preferred
Arg-Gly-Asp-Ser (RGDS) is a tetrapeptide containing the integrin-binding RGD sequence used as an integrin inhibitor and analytical standard for research on cell adhesion, integrin signaling, and extracellular matrix interactions. Supplied as a white to off-white solid, it is suitable for biochemical and cell biology assays.
  • Tetrapeptide containing the RGD integrin-binding motif.
  • Used as an integrin inhibitor and in cell adhesion assays.
  • High reported purity (LCMS 99.09%).
  • Molecular weight 433.42 g/mol and formula C15H27N7O8.
  • White to off-white solid suitable for analytical use.
Catalog No. 50-002-30960 Supplier Medchemexpress LLC Supplier No. HY12290100MG
Arg-Gly-Asp-Ser (RGDS) is a tetrapeptide containing the integrin-binding RGD sequence used as an integrin inhibitor and analytical standard for research on cell adhesion, integrin signaling, and extracellular matrix interactions. Supplied as a white to off-white solid, it is suitable for biochemical and cell biology assays.
  • Tetrapeptide containing the RGD integrin-binding motif.
  • Used as an integrin inhibitor and in cell adhesion assays.
  • High reported purity (LCMS 99.09%).
  • Molecular weight 433.42 g/mol and formula C15H27N7O8.
  • White to off-white solid suitable for analytical use.