Learn More
TARGETMOL CHEMICALS INC Argipressin acetate 113-79-1fr
SDP
Catalog No. 502880138
Encompass_Preferred
Also available in 1 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Vasopressin acetate is a peptide hormone with vasoconstrictive and antidiuretic activities that binds to the vascular arginine vasopressin receptor, V1, with Kd values of 1.31 and 1.44 nM in A7r5 rat aortic smooth muscle cells and neonatal rat cardiomyocytes, respectively Purity 100%
Catalog No. 50-288-0138 Supplier TARGETMOL CHEMICALS INC Supplier No. T728425MG
Add to Cart
Add to Cart
Also available in 1 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Vasopressin acetate is a peptide hormone with vasoconstrictive and antidiuretic activities that binds to the vascular arginine vasopressin receptor, V1, with Kd values of 1.31 and 1.44 nM in A7r5 rat aortic smooth muscle cells and neonatal rat cardiomyocytes, respectively Purity 100%