Learn More
Apexbio Technology LLC Argipressin acetate 129979-57-3 1mg
SDP
Catalog No. 5000095906
Encompass_Preferred
Argipressin acetate (CAS 129979-57-3) is a synthetic peptide analog of the antidiuretic hormone vasopressin formulated as the acetate salt It selectively binds to renal V2 receptors enhancing water reabsorption in the collecting ducts and thereby reducing urine output Additionally Argipressin acetate activates vascular V1 receptors on smooth muscle cells resulting in vasoconstriction and increased blood pressure This dual activity underlies its use in research focused on hemostasis vascular tone regulation and the study of water-electrolyte balance as well as in models of antidiuretic hormone deficiency and acute hemorrhagic conditions
Catalog No. 50-000-95906 Supplier Apexbio Technology LLC Supplier No. A99311
Argipressin acetate (CAS 129979-57-3) is a synthetic peptide analog of the antidiuretic hormone vasopressin formulated as the acetate salt It selectively binds to renal V2 receptors enhancing water reabsorption in the collecting ducts and thereby reducing urine output Additionally Argipressin acetate activates vascular V1 receptors on smooth muscle cells resulting in vasoconstriction and increased blood pressure This dual activity underlies its use in research focused on hemostasis vascular tone regulation and the study of water-electrolyte balance as well as in models of antidiuretic hormone deficiency and acute hemorrhagic conditions