Learn More
AdipoGen Arglabin (5 mg)
SDP
Supplier:  AdipoGen AGCN20458M005
Encompass_Preferred
Chemical. CAS: 84692-91-1. Formula: C15H18O3. MW: 246.3. Semi-synthetic from parthenolide (AG-CN2-0455), which is isolated from Magnolia grandiflora. Anticancer compound. Farnesyltransferase inhibitor. Potent anti-inflammatory agent. NF-kappaB inhibitor. Inflammasome inhibitor. Antiatherogenic. Prevents the accumulation of lipid containing plaques in arteries.
Catalog No. 50-205-3630