Learn More
Medchemexpress LLC HY-16059 5mg Medchemexpress, Arglabin CAS:84692-91-1 Purity:>98%
SDP
Catalog No. 502081867
Encompass_Preferred
Medchemexpress, HY-16059 5mg Arglabin CAS:84692-91-1 Arglabin ((+)-Arglabin), a natural product isolated from Artemisia glabella, is a NLRP3 inflammasome inhibitor. Arglabin shows anti-inflammatory and antitumor activities. The antitumor activity of Arglabin proceeds through its inhibition of farnesyl transferase which leads to the activation of RAS proto-oncogene. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1867 Supplier Medchemexpress LLC Supplier No. HY160595MG
Medchemexpress, HY-16059 5mg Arglabin CAS:84692-91-1 Arglabin ((+)-Arglabin), a natural product isolated from Artemisia glabella, is a NLRP3 inflammasome inhibitor. Arglabin shows anti-inflammatory and antitumor activities. The antitumor activity of Arglabin proceeds through its inhibition of farnesyl transferase which leads to the activation of RAS proto-oncogene. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.