Learn More
GENERAL BIOPHYSICS LLC Argon Gas Analyzer Model A
Catalog No. NC3361679
Encompass
Thermoconductivity Gas Analyzer for Argon with Onboard PumpApplication: Calibrated for argon gas with onboard pump to take gas samples.Features: Includes data display screen.Accuracy: ±0.9%.
Catalog No. NC3361679 Supplier GENERAL BIOPHYSICS LLC Supplier No. GAARO2A
Add to Cart
Add to Cart
Request a Quote
Thermoconductivity Gas Analyzer for Argon with Onboard PumpApplication: Calibrated for argon gas with onboard pump to take gas samples.Features: Includes data display screen.Accuracy: ±0.9%.