Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Argonaute-2 inhibitor BCI-137 | 112170-24-8 | MFCD05148320 | 99.6% | 313.29 g/mol | C11H11N3O6S | 10 MM 1 ML
SDP

Catalog No. 5000307542
Encompass_Preferred

BCI-137 is a small-molecule competitive inhibitor of Argonaute 2 (AGO2) used to disrupt AGO2-miRNA interactions in biochemical and cellular research. It is supplied as a 10 mM solution in DMSO (1 mL).

  • Inhibits AGO2 by competing with miRNA binding.
  • Reported IC50 for AGO2 competition: 342 μM.
  • Supplied as a 10 mM solution in DMSO, 1 mL volume.
  • Molecular weight 313.29 g/mol; formula C11H11N3O6S.
  • High purity (manufacturer listing ≈99.6%).
  • Soluble in DMSO (100 mg/mL); example in vivo vehicle recipes provided.
  • Recommended storage: powder -20°C; solutions -80°C.

Catalog No. 50-003-07542 Supplier Medchemexpress LLC Supplier No. HY11902410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BCI-137 is a small-molecule competitive inhibitor of Argonaute 2 (AGO2) used to disrupt AGO2-miRNA interactions in biochemical and cellular research. It is supplied as a 10 mM solution in DMSO (1 mL).

  • Inhibits AGO2 by competing with miRNA binding.
  • Reported IC50 for AGO2 competition: 342 μM.
  • Supplied as a 10 mM solution in DMSO, 1 mL volume.
  • Molecular weight 313.29 g/mol; formula C11H11N3O6S.
  • High purity (manufacturer listing ≈99.6%).
  • Soluble in DMSO (100 mg/mL); example in vivo vehicle recipes provided.
  • Recommended storage: powder -20°C; solutions -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.