Learn More
Sino Biological Recombinant Human Argonaute-2/AGO2 Protein (His Tag)
Supplier:  Sino Biological 11079H07B100
Encompass_Preferred
A DNA sequence encoding the full length of human AGO2 (NP_036286.2) (Met 1-Ala 859) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-211-6291