Learn More
Cole-Parmer Argos Technologies PolarSafe Cardboard Freezer Box, 5-3/4" x 5-3/4" x 4-7/8"H; without Divider
Catalog No. NC1677382 Shop All Cole-Parmer Products
Encompass_Preferred
These economical cryo/freezer storage boxes offer excellent low-temperature storage. Moisture-repellent coating protects boxes from effects of liquid nitrogen or condensation.
Catalog No. NC1677382 Supplier Cole-Parmer Supplier No. 0675575
These economical cryo/freezer storage boxes offer excellent low-temperature storage. Moisture-repellent coating protects boxes from effects of liquid nitrogen or condensation.