Learn More
Cole-Parmer Argos Technologies Disposable Serological Pipettes, 100 mL, Individually Wrap; 50/Cs
Catalog No. NC1574404 Shop All Cole-Parmer Products
Encompass_Preferred
Designed for accurate and rapid fluid dispensing, these disposable polystyrene (PS) serological pipettes feature a filter plug which eliminates lint fibers and a uniform mouth piece. Sharp, black, permanent graduations guarantee precise reading and accurate dispensing to ±2%. Reverse graduations show volume removed or remaining on all pipettes larger than 1 mL. Negative graduations provide additional pipetting volume.
Pipette with 1.1-mL capacity meets the APHA standards for the examination of dairy products.
The pipettes come individually wrapped in a fiber-free paper with easy peel-apart opening. Color-coded dispenser box allows quick access to pipettes without opening top.
Catalog No. NC1574404 Supplier Cole-Parmer Supplier No. 0439522
Designed for accurate and rapid fluid dispensing, these disposable polystyrene (PS) serological pipettes feature a filter plug which eliminates lint fibers and a uniform mouth piece. Sharp, black, permanent graduations guarantee precise reading and accurate dispensing to ±2%. Reverse graduations show volume removed or remaining on all pipettes larger than 1 mL. Negative graduations provide additional pipetting volume.
Pipette with 1.1-mL capacity meets the APHA standards for the examination of dairy products.
The pipettes come individually wrapped in a fiber-free paper with easy peel-apart opening. Color-coded dispenser box allows quick access to pipettes without opening top.