Learn More
Cole-Parmer Argos Technologies Pestle for 1.5 mL Microtube, 100/pk
Supplier:  Cole-Parmer 4446819
Encompass_Preferred
Plastic pestles and tubes are designed to homogenize cells and plant tissue or to resuspend protein and DNA pellets. They are DNAase/RNAase and pyrogen free, and come individually wrapped to guarantee purity.
Catalog No. NC0782485