Learn More
Cole-Parmer Argos Technologies Vortamix™ Mini Vortexer, 100-240 VAC; US Adapter
Catalog No. NC1777542 Shop All Cole-Parmer Products
Encompass_Preferred
Compact vortex shaker mixes samples in centrifuge and test tubes, up to 30 mm in diameter, with little effort. The vortex motion is activated by placing a tube on the gasket cap and pressing down lightly.
Catalog No. NC1777542 Supplier Cole-Parmer Supplier No. 0439727
Compact vortex shaker mixes samples in centrifuge and test tubes, up to 30 mm in diameter, with little effort. The vortex motion is activated by placing a tube on the gasket cap and pressing down lightly.