Learn More
Cole-Parmer Pipette Basins, 12 Channel x 3 mL, Non-Sterile, Bulk; 50/cs
Catalog No. NC1678038 Shop All Cole-Parmer Products
Encompass_Preferred
The pipette reagent reservoirs feature a V-shaped bottom, making multichannel pipetting easier and ensuring complete aspiration of costly reagents. They are available in a variety of volumes come in either sterile or nonsterile formats.
Catalog No. NC1678038 Supplier Cole-Parmer Supplier No. 0439532
Add to Cart
Add to Cart
The pipette reagent reservoirs feature a V-shaped bottom, making multichannel pipetting easier and ensuring complete aspiration of costly reagents. They are available in a variety of volumes come in either sterile or nonsterile formats.