Learn More
ATCC ARH-77; Plasma Cell Leukemia; Human (Homo sapiens)
Catalog No. 502383319
Encompass
ARH-77 is a B lymphoblast cell line that was isolated from the peripheral blood of a White, 33-year-old, female patient with plasma cell leukemia. This cell line was deposited by BD Drewinko and can be used in immunology and immune system disorder research.
Catalog No. 50-238-3319 Supplier ATCC Supplier No. CRL1621
ARH-77 is a B lymphoblast cell line that was isolated from the peripheral blood of a White, 33-year-old, female patient with plasma cell leukemia. This cell line was deposited by BD Drewinko and can be used in immunology and immune system disorder research.