Learn More
Aviva Systems Biology Corp ARHGAP1 Antibody - N-terminal region (ARP54722_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54722_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARHGAP1. It was validated on Western Blot. ARHGAP1,Rho GTPase activating protein 1,RHOGAP; RHOGAP1; CDC42GAP; p50rhoGAP; ARHGAP1,rho GTPase-activating protein 1,rho GTPase-activating protein 1,p50-RhoGAP,CDC42 GTPase-activating protein,GTPase-activating protein rhoOGAP,rho-type GTPase-activating protein 1,rho-related small GTPase protein activatorThe immunogen is a synthetic peptide directed towards the N terminal region of human ARHGAP1. Alternate names: CDC42GAP, RHOGAP, RHOGAP1, p50rhoGAPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-961