Learn More
Aviva Systems Biology Corp ARHGAP15 Antibody - middle region (ARP57261_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57261_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARHGAP15. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ARHGAP15. Alternate names: BM046ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-287