Learn More
Aviva Systems Biology Corp ARHGAP19 Antibody - C-terminal region (ARP60716_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60716_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARHGAP19. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ARHGAP19. Alternate names: DKFZp313K217, MGC138804, MGC138805, MGC14258ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-141