Learn More
Aviva Systems Biology Corp ARHGAP20 Antibody - middle region (ARP57460_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57460_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARHGAP20. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ARHGAP20. Alternate names: KIAA1391, RARHOGAPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-316