Learn More
Aviva Systems Biology Corp ARHGAP25 Antibody - middle region (ARP58423_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58423_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARHGAP25. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ARHGAP25. Alternate names: KAIA0053ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-62303