Learn More
Aviva Systems Biology Corp ARHGAP28 Antibody - C-terminal region (ARP53571_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53571_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARHGAP28. It was validated on Western Blot. . Alternate names: DKFZp686A2038, FLJ10312ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-852