Learn More
Aviva Systems Biology Corp ARHGAP8 Antibody - middle region (ARP65532_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65532_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARHGAP8. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Human ARHGAP8. Alternate names: BPGAP1, FLJ20185, PP610ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-288-513